Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFN1
Entrez Gene ID
55669
Gene Name
mitofusin 1
Gene Aliases
hfzo1, hfzo2
Chromosome Location
Chr.3: 179347692 - 179394944 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033540.2 NP_284941.2 17 - 18 2267 151
XM_005247596.3 XP_005247653.2 17 - 18 2225 151
XM_011512963.2 XP_011511265.1 14 - 15 1853 151
GenBank mRNA AB043588.1 - 16 - 17 2232 151
AB043589.1 - 17 - 18 2232 151
AF054986.1 - 7 - 8 1024 151
AF329637.1 - 16 - 17 2141 151
AK000700.1 - 16 - 17 2097 151
AK314306.1 - 17 - 18 2224 151
BC040557.1 - 17 - 18 2267 151
CR627379.1 - 14 - 15 1916 151
U95822.1 - 11 - 12 1397 151

View Full Product Details