Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFN1
Entrez Gene ID
55669
Gene Name
mitofusin 1
Gene Aliases
hfzo1, hfzo2
Chromosome Location
Chr.3: 179347692 - 179394944 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK000700.1 - 10 - 11 1183 77

View Full Product Details