Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNAPC4
Entrez Gene ID
6621
Gene Name
small nuclear RNA activating complex polypeptide 4
Gene Aliases
PTFalpha, SNAP190
Chromosome Location
Chr.9: 136375571 - 136400220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003086.2 NP_003077.2 14 - 15 1513 85
XM_005266096.2 XP_005266153.1 15 - 16 1604 85
XM_006717241.2 XP_006717304.1 15 - 16 1588 85
XM_006717242.3 XP_006717305.1 15 - 16 2517 85
XM_017015058.1 XP_016870547.1 14 - 15 3033 85
GenBank mRNA AF032387.1 - 14 - 15 1879 85

View Full Product Details