The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003086.2 | NP_003077.2 | 19 - 20 | 2512 | 131 |
XM_005266096.2 | XP_005266153.1 | 20 - 21 | 2603 | 131 | |
XM_006717241.2 | XP_006717304.1 | 20 - 21 | 2587 | 131 | |
XM_006717242.3 | XP_006717305.1 | 20 - 21 | 3516 | 131 | |
XM_006717244.2 | XP_006717307.1 | 5 - 6 | 954 | 131 | |
XM_017015058.1 | XP_016870547.1 | 19 - 20 | 3948 | 131 | |
GenBank mRNA | AF032387.1 | - | 19 - 20 | 2878 | 131 |