Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNAPC4
Entrez Gene ID
6621
Gene Name
small nuclear RNA activating complex polypeptide 4
Gene Aliases
PTFalpha, SNAP190
Chromosome Location
Chr.9: 136375571 - 136400220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003086.2 NP_003077.2 8 - 9 820 76
XM_005266096.2 XP_005266153.1 9 - 10 911 76
XM_006717241.2 XP_006717304.1 9 - 10 895 76
XM_006717242.3 XP_006717305.1 9 - 10 1824 76
XM_017015058.1 XP_016870547.1 8 - 9 2340 76
GenBank mRNA AF032387.1 - 8 - 9 1186 76

View Full Product Details