Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNAPC4
Entrez Gene ID
6621
Gene Name
small nuclear RNA activating complex polypeptide 4
Gene Aliases
PTFalpha, SNAP190
Chromosome Location
Chr.9: 136375571 - 136400220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003086.2 NP_003077.2 22 - 23 4433 100
XM_005266096.2 XP_005266153.1 23 - 24 4524 100
XM_006717241.2 XP_006717304.1 23 - 24 4508 100
XM_006717242.3 XP_006717305.1 23 - 24 5437 100
XM_006717244.2 XP_006717307.1 8 - 9 2875 100
XM_017015058.1 XP_016870547.1 22 - 23 5869 100
GenBank mRNA AF032387.1 - 22 - 23 4799 100

View Full Product Details