The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308330.1 | NP_001295259.1 | 3 - 4 | 432 | 72 |
NM_014980.2 | NP_055795.1 | 3 - 4 | 432 | 72 | |
XM_006713825.3 | XP_006713888.1 | 3 - 4 | 602 | 72 | |
XM_011513328.2 | XP_011511630.1 | 2 - 3 | 279 | 72 | |
XM_011513331.2 | XP_011511633.1 | 3 - 4 | 429 | 72 | |
XM_011513333.2 | XP_011511635.1 | 3 - 4 | 429 | 72 | |
XM_017007530.1 | XP_016863019.1 | 3 - 4 | 363 | 72 | |
XM_017007531.1 | XP_016863020.1 | 3 - 4 | 417 | 72 | |
XM_017007532.1 | XP_016863021.1 | 3 - 4 | 366 | 72 | |
XM_017007534.1 | XP_016863023.1 | 3 - 4 | 429 | 72 | |
XM_017007535.1 | XP_016863024.1 | 3 - 4 | 620 | 72 | |
GenBank mRNA | AB023223.1 | - | 3 - 4 | 399 | 72 |
AK296544.1 | - | 3 - 4 | 430 | 72 | |
BC022029.1 | - | 3 - 4 | 410 | 72 | |
DC324331.1 | - | 3 - 4 | 432 | 72 |