The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308330.1 | NP_001295259.1 | 5 - 6 | 609 | 108 |
NM_014980.2 | NP_055795.1 | 5 - 6 | 609 | 108 | |
XM_006713825.3 | XP_006713888.1 | 5 - 6 | 779 | 108 | |
XM_011513328.2 | XP_011511630.1 | 4 - 5 | 456 | 108 | |
XM_011513331.2 | XP_011511633.1 | 5 - 6 | 606 | 108 | |
XM_011513333.2 | XP_011511635.1 | 5 - 6 | 606 | 108 | |
XM_017007530.1 | XP_016863019.1 | 5 - 6 | 540 | 108 | |
XM_017007531.1 | XP_016863020.1 | 5 - 6 | 594 | 108 | |
XM_017007532.1 | XP_016863021.1 | 5 - 6 | 543 | 108 | |
XM_017007533.1 | XP_016863022.1 | 1 - 2 | 100 | 108 | |
XM_017007534.1 | XP_016863023.1 | 5 - 6 | 606 | 108 | |
XM_017007535.1 | XP_016863024.1 | 5 - 6 | 797 | 108 | |
GenBank mRNA | AB023223.1 | - | 5 - 6 | 576 | 108 |
AK296544.1 | - | 5 - 6 | 607 | 108 | |
BC022029.1 | - | 5 - 6 | 587 | 108 |