The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022828.3 | NP_073739.3 | 2 - 3 | 473 | 98 |
XM_011543581.1 | XP_011541883.1 | 2 - 3 | 484 | 98 | |
XM_011543582.2 | XP_011541884.1 | 2 - 3 | 458 | 98 | |
XM_011543586.2 | XP_011541888.1 | 2 - 3 | 458 | 98 | |
XM_017009730.1 | XP_016865219.1 | 2 - 3 | 484 | 98 | |
GenBank mRNA | AK310855.1 | - | 2 - 3 | 458 | 98 |
BC137285.1 | - | 2 - 3 | 423 | 98 | |
CA943109.1 | - | 2 - 3 | 457 | 98 |