The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022828.3 | NP_073739.3 | 18 - 19 | 2522 | 114 |
XM_011543581.1 | XP_011541883.1 | 18 - 19 | 2533 | 114 | |
XM_011543582.2 | XP_011541884.1 | 18 - 19 | 2507 | 114 | |
XM_011543586.2 | XP_011541888.1 | 18 - 19 | 2507 | 114 | |
XM_017009730.1 | XP_016865219.1 | 18 - 19 | 2533 | 114 | |
XM_017009731.1 | XP_016865220.1 | 17 - 18 | 2308 | 114 | |
XM_017009732.1 | XP_016865221.1 | 17 - 18 | 2308 | 114 | |
XM_017009733.1 | XP_016865222.1 | 16 - 17 | 2136 | 114 | |
XM_017009734.1 | XP_016865223.1 | 16 - 17 | 2136 | 114 | |
XM_017009735.1 | XP_016865224.1 | 15 - 16 | 1955 | 114 | |
GenBank mRNA | AK025593.1 | - | 2 - 3 | 135 | 114 |
AK131450.1 | - | 17 - 18 | 2215 | 114 | |
AK225316.1 | - | 2 - 3 | 135 | 114 | |
BC137285.1 | - | 18 - 19 | 2472 | 114 | |
BM827416.1 | - | 4 - 5 | 303 | 114 |