The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190482.1 | NP_001177411.1 | 3 - 4 | 947 | 56 |
NM_006200.5 | NP_006191.2 | 3 - 4 | 947 | 56 | |
NR_120409.1 | 3 - 4 | 947 | 56 | ||
XM_005252039.3 | XP_005252096.1 | 3 - 4 | 990 | 56 | |
XM_011518769.2 | XP_011517071.1 | 3 - 4 | 990 | 56 | |
GenBank mRNA | AK056202.1 | - | 3 - 4 | 921 | 56 |
AK225773.1 | - | 3 - 4 | 944 | 56 | |
AK310862.1 | - | 3 - 4 | 915 | 56 | |
AL834522.1 | - | 3 - 4 | 477 | 56 | |
BC012064.1 | - | 3 - 4 | 886 | 56 | |
U49114.1 | - | 3 - 4 | 367 | 56 | |
U56387.2 | - | 3 - 4 | 421 | 56 |