Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCSK5
Entrez Gene ID
5125
Gene Name
proprotein convertase subtilisin/kexin type 5
Gene Aliases
PC5, PC6, PC6A, SPC6
Chromosome Location
Chr.9: 75890601 - 76362339 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190482.1 NP_001177411.1 8 - 9 1650 89
NM_006200.5 NP_006191.2 8 - 9 1650 89
NR_120409.1 8 - 9 1650 89
XM_005252039.3 XP_005252096.1 8 - 9 1693 89
XM_011518769.2 XP_011517071.1 8 - 9 1693 89
XM_017014800.1 XP_016870289.1 1 - 2 70 89
GenBank mRNA AK056202.1 - 8 - 9 1624 89
AK225773.1 - 8 - 9 1647 89
AL834522.1 - 8 - 9 1180 89
BC012064.1 - 8 - 9 1589 89
U49114.1 - 8 - 9 1070 89
U56387.2 - 8 - 9 1124 89

View Full Product Details