Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RXRB
Entrez Gene ID
6257
Gene Name
retinoid X receptor beta
Gene Aliases
DAUDI6, H-2RIIBP, NR2B2, RCoR-1
Chromosome Location
Chr.6: 33193585 - 33200997 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270401.1 NP_001257330.1 2 - 3 701 55
NM_021976.4 NP_068811.1 2 - 3 701 55
XM_017011176.1 XP_016866665.1 1 - 2 290 55
GenBank mRNA AB209244.1 - 2 - 3 619 55
AK225842.1 - 2 - 3 660 55
AK300402.1 - 2 - 3 677 55
BC001167.1 - 2 - 3 660 55
BT007280.1 - 2 - 3 481 55
HQ692844.1 - 2 - 3 481 55
HQ709179.1 - 2 - 3 481 55
KU178313.1 - 2 - 3 481 55
KU178314.1 - 2 - 3 481 55
KU178315.1 - 2 - 3 481 55
M84820.1 - 2 - 3 598 55
X63522.1 - 2 - 3 660 55

View Full Product Details