The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024855.3 | NP_079131.3 | 3 - 4 | 803 | 108 |
GenBank mRNA | AK022847.1 | - | 3 - 4 | 788 | 108 |
BC001324.1 | - | 2 - 3 | 202 | 108 | |
BC038402.1 | - | 3 - 4 | 803 | 108 |