Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMP10
Entrez Gene ID
4319
Gene Name
matrix metallopeptidase 10
Gene Aliases
SL-2, STMY2
Chromosome Location
Chr.11: 102770502 - 102780628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002425.2 NP_002416.1 8 - 9 1265 75
GenBank mRNA AK222601.1 - 8 - 9 1265 75
AK313960.1 - 8 - 9 1265 75
BC002591.2 - 8 - 9 1252 75
BT007442.1 - 8 - 9 1228 75
X07820.1 - 8 - 9 1250 75

View Full Product Details