The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020872.2 | NP_065923.1 | 5 - 6 | 682 | 97 |
XM_005264757.3 | XP_005264814.1 | 6 - 7 | 792 | 97 | |
XM_011533768.2 | XP_011532070.1 | 5 - 6 | 4314 | 97 | |
XM_017006507.1 | XP_016861996.1 | 6 - 7 | 1362 | 97 | |
XM_017006508.1 | XP_016861997.1 | 5 - 6 | 587 | 97 | |
GenBank mRNA | AB040929.1 | - | 3 - 4 | 331 | 97 |
BC150608.1 | - | 5 - 6 | 682 | 97 |