The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020872.2 | NP_065923.1 | 9 - 10 | 1239 | 134 |
XM_005264757.3 | XP_005264814.1 | 10 - 11 | 1349 | 134 | |
XM_011533768.2 | XP_011532070.1 | 9 - 10 | 4871 | 134 | |
XM_017006507.1 | XP_016861996.1 | 10 - 11 | 1919 | 134 | |
XM_017006508.1 | XP_016861997.1 | 9 - 10 | 1144 | 134 | |
GenBank mRNA | AB040929.1 | - | 7 - 8 | 888 | 134 |
BC150608.1 | - | 9 - 10 | 1239 | 134 |