The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172477.1 | NP_001165948.1 | 6 - 7 | 907 | 79 |
NM_001172478.1 | NP_001165949.1 | 5 - 6 | 770 | 79 | |
NM_015713.4 | NP_056528.2 | 6 - 7 | 926 | 79 | |
GenBank mRNA | AB036063.1 | - | 6 - 7 | 926 | 79 |
AB163437.1 | - | 5 - 6 | 526 | 79 | |
AB166671.1 | - | 6 - 7 | 682 | 79 | |
AK001965.1 | - | 6 - 7 | 721 | 79 | |
AK304354.1 | - | 6 - 7 | 851 | 79 | |
AL137348.1 | - | 6 - 7 | 728 | 79 | |
BC042468.1 | - | 4 - 5 | 484 | 79 | |
BC108261.1 | - | 6 - 7 | 738 | 79 | |
BC117496.1 | - | 6 - 7 | 763 | 79 | |
BC130628.1 | - | 6 - 7 | 763 | 79 | |
CR627376.1 | - | 6 - 7 | 851 | 79 |