The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006076.4 | NP_006067.3 | 2 - 3 | 451 | 73 |
XM_005250306.2 | XP_005250363.1 | 2 - 3 | 437 | 73 | |
XM_017012112.1 | XP_016867601.1 | 2 - 3 | 392 | 73 | |
XM_017012113.1 | XP_016867602.1 | 2 - 3 | 437 | 73 | |
XM_017012114.1 | XP_016867603.1 | 2 - 3 | 437 | 73 | |
GenBank mRNA | AK314954.1 | - | 2 - 3 | 437 | 73 |
BC009393.2 | - | 2 - 3 | 450 | 73 | |
BC017329.2 | - | 2 - 3 | 403 | 73 | |
BF314987.1 | - | 2 - 3 | 451 | 73 |