Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGFG2
Entrez Gene ID
3268
Gene Name
ArfGAP with FG repeats 2
Gene Aliases
HRBL, RABR
Chromosome Location
Chr.7: 100539211 - 100568220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006076.4 NP_006067.3 3 - 4 568 89
XM_005250306.2 XP_005250363.1 3 - 4 554 89
XM_017012112.1 XP_016867601.1 3 - 4 509 89
XM_017012113.1 XP_016867602.1 3 - 4 554 89
GenBank mRNA AF015042.1 - 3 - 4 492 89
AK314954.1 - 3 - 4 554 89
BC017329.2 - 3 - 4 520 89

View Full Product Details