Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM22
Entrez Gene ID
53616
Gene Name
ADAM metallopeptidase domain 22
Gene Aliases
ADAM 22, MDC2
Chromosome Location
Chr.7: 87934143 - 88202889 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324417.1 NP_001311346.1 14 - 15 1539 99
NM_001324418.1 NP_001311347.1 14 - 15 1542 99
NM_001324419.1 NP_001311348.1 14 - 15 1539 99
NM_001324420.1 NP_001311349.1 14 - 15 1542 99
NM_001324421.1 NP_001311350.1 14 - 15 1542 99
NM_004194.4 NP_004185.1 14 - 15 1542 99
NM_016351.5 NP_057435.2 14 - 15 1542 99
NM_021721.4 NP_068367.1 14 - 15 1542 99
NM_021722.5 NP_068368.2 14 - 15 1542 99
NM_021723.4 NP_068369.1 14 - 15 1542 99
XM_005250445.3 XP_005250502.1 13 - 14 1390 99
XM_006716028.3 XP_006716091.1 13 - 14 1390 99
XM_006716029.3 XP_006716092.1 13 - 14 1390 99
XM_011516318.1 XP_011514620.1 14 - 15 1434 99
XM_011516319.2 XP_011514621.1 14 - 15 1518 99
XM_011516320.2 XP_011514622.1 13 - 14 1390 99
XM_011516321.2 XP_011514623.1 13 - 14 1390 99
XM_011516322.1 XP_011514624.1 14 - 15 1434 99
XM_011516323.1 XP_011514625.1 14 - 15 1434 99
XM_011516324.1 XP_011514626.1 14 - 15 1106 99
XM_017012329.1 XP_016867818.1 15 - 16 1485 99
XM_017012330.1 XP_016867819.1 15 - 16 1485 99
XM_017012331.1 XP_016867820.1 14 - 15 1515 99
XM_017012332.1 XP_016867821.1 14 - 15 1434 99
XM_017012333.1 XP_016867822.1 14 - 15 1434 99
XM_017012334.1 XP_016867823.1 15 - 16 1485 99
XM_017012335.1 XP_016867824.1 15 - 16 1485 99
XM_017012336.1 XP_016867825.1 14 - 15 1434 99
XM_017012337.1 XP_016867826.1 15 - 16 1485 99
XM_017012338.1 XP_016867827.1 14 - 15 1513 99
XM_017012339.1 XP_016867828.1 15 - 16 1485 99
XM_017012340.1 XP_016867829.1 3 - 4 165 99
XM_017012341.1 XP_016867830.1 13 - 14 1162 99
GenBank mRNA AB009671.1 - 14 - 15 1340 99
AF073291.2 - 14 - 15 1265 99
AF155381.1 - 14 - 15 1265 99
AF155382.1 - 14 - 15 1265 99
AF158637.1 - 13 - 14 1102 99
AK125432.1 - 13 - 14 1405 99
AK291010.1 - 14 - 15 1389 99
BC125118.1 - 14 - 15 1296 99

View Full Product Details