Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM22
Entrez Gene ID
53616
Gene Name
ADAM metallopeptidase domain 22
Gene Aliases
ADAM 22, MDC2
Chromosome Location
Chr.7: 87934143 - 88202889 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324417.1 NP_001311346.1 16 - 17 1716 97
NM_001324418.1 NP_001311347.1 16 - 17 1719 97
NM_001324419.1 NP_001311348.1 16 - 17 1716 97
NM_001324420.1 NP_001311349.1 16 - 17 1719 97
NM_001324421.1 NP_001311350.1 16 - 17 1719 97
NM_004194.4 NP_004185.1 16 - 17 1719 97
NM_016351.5 NP_057435.2 16 - 17 1719 97
NM_021721.4 NP_068367.1 16 - 17 1719 97
NM_021722.5 NP_068368.2 16 - 17 1719 97
NM_021723.4 NP_068369.1 16 - 17 1719 97
XM_005250445.3 XP_005250502.1 15 - 16 1567 97
XM_006716028.3 XP_006716091.1 15 - 16 1567 97
XM_006716029.3 XP_006716092.1 15 - 16 1567 97
XM_011516318.1 XP_011514620.1 16 - 17 1611 97
XM_011516319.2 XP_011514621.1 16 - 17 1695 97
XM_011516320.2 XP_011514622.1 15 - 16 1567 97
XM_011516321.2 XP_011514623.1 15 - 16 1567 97
XM_011516322.1 XP_011514624.1 16 - 17 1611 97
XM_011516323.1 XP_011514625.1 16 - 17 1611 97
XM_011516324.1 XP_011514626.1 16 - 17 1283 97
XM_017012329.1 XP_016867818.1 17 - 18 1662 97
XM_017012330.1 XP_016867819.1 17 - 18 1662 97
XM_017012331.1 XP_016867820.1 16 - 17 1692 97
XM_017012332.1 XP_016867821.1 16 - 17 1611 97
XM_017012333.1 XP_016867822.1 16 - 17 1611 97
XM_017012334.1 XP_016867823.1 17 - 18 1662 97
XM_017012335.1 XP_016867824.1 17 - 18 1662 97
XM_017012336.1 XP_016867825.1 16 - 17 1611 97
XM_017012337.1 XP_016867826.1 17 - 18 1662 97
XM_017012338.1 XP_016867827.1 16 - 17 1690 97
XM_017012339.1 XP_016867828.1 17 - 18 1662 97
XM_017012340.1 XP_016867829.1 5 - 6 342 97
XM_017012341.1 XP_016867830.1 15 - 16 1339 97
GenBank mRNA AB009671.1 - 16 - 17 1517 97
AF073291.2 - 16 - 17 1442 97
AF155381.1 - 16 - 17 1442 97
AF155382.1 - 16 - 17 1442 97
AF158637.1 - 15 - 16 1279 97
AK125432.1 - 15 - 16 1582 97
AK291010.1 - 16 - 17 1566 97
BC125118.1 - 16 - 17 1473 97

View Full Product Details