Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM22
Entrez Gene ID
53616
Gene Name
ADAM metallopeptidase domain 22
Gene Aliases
ADAM 22, MDC2
Chromosome Location
Chr.7: 87934143 - 88202889 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324417.1 NP_001311346.1 17 - 18 1806 64
NM_001324418.1 NP_001311347.1 17 - 18 1809 64
NM_001324419.1 NP_001311348.1 17 - 18 1806 64
NM_001324420.1 NP_001311349.1 17 - 18 1809 64
NM_001324421.1 NP_001311350.1 17 - 18 1809 64
NM_004194.4 NP_004185.1 17 - 18 1809 64
NM_016351.5 NP_057435.2 17 - 18 1809 64
NM_021721.4 NP_068367.1 17 - 18 1809 64
NM_021722.5 NP_068368.2 17 - 18 1809 64
NM_021723.4 NP_068369.1 17 - 18 1809 64
XM_005250445.3 XP_005250502.1 16 - 17 1657 64
XM_006716028.3 XP_006716091.1 16 - 17 1657 64
XM_006716029.3 XP_006716092.1 16 - 17 1657 64
XM_011516318.1 XP_011514620.1 17 - 18 1701 64
XM_011516319.2 XP_011514621.1 17 - 18 1785 64
XM_011516320.2 XP_011514622.1 16 - 17 1657 64
XM_011516321.2 XP_011514623.1 16 - 17 1657 64
XM_011516322.1 XP_011514624.1 17 - 18 1701 64
XM_011516323.1 XP_011514625.1 17 - 18 1701 64
XM_011516324.1 XP_011514626.1 17 - 18 1373 64
XM_017012329.1 XP_016867818.1 18 - 19 1752 64
XM_017012330.1 XP_016867819.1 18 - 19 1752 64
XM_017012331.1 XP_016867820.1 17 - 18 1782 64
XM_017012332.1 XP_016867821.1 17 - 18 1701 64
XM_017012333.1 XP_016867822.1 17 - 18 1701 64
XM_017012334.1 XP_016867823.1 18 - 19 1752 64
XM_017012335.1 XP_016867824.1 18 - 19 1752 64
XM_017012336.1 XP_016867825.1 17 - 18 1701 64
XM_017012337.1 XP_016867826.1 18 - 19 1752 64
XM_017012338.1 XP_016867827.1 17 - 18 1780 64
XM_017012339.1 XP_016867828.1 18 - 19 1752 64
XM_017012340.1 XP_016867829.1 6 - 7 432 64
XM_017012341.1 XP_016867830.1 16 - 17 1429 64
GenBank mRNA AB009671.1 - 17 - 18 1607 64
AF073291.2 - 17 - 18 1532 64
AF155381.1 - 17 - 18 1532 64
AF155382.1 - 17 - 18 1532 64
AF158637.1 - 16 - 17 1369 64
AK125432.1 - 16 - 17 1672 64
AK291010.1 - 17 - 18 1656 64
BC125118.1 - 17 - 18 1563 64

View Full Product Details