Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM22
Entrez Gene ID
53616
Gene Name
ADAM metallopeptidase domain 22
Gene Aliases
ADAM 22, MDC2
Chromosome Location
Chr.7: 87934143 - 88202889 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324417.1 NP_001311346.1 8 - 9 997 145
NM_001324418.1 NP_001311347.1 8 - 9 1000 145
NM_001324419.1 NP_001311348.1 8 - 9 997 145
NM_001324420.1 NP_001311349.1 8 - 9 1000 145
NM_001324421.1 NP_001311350.1 8 - 9 1000 145
NM_004194.4 NP_004185.1 8 - 9 1000 145
NM_016351.5 NP_057435.2 8 - 9 1000 145
NM_021721.4 NP_068367.1 8 - 9 1000 145
NM_021722.5 NP_068368.2 8 - 9 1000 145
NM_021723.4 NP_068369.1 8 - 9 1000 145
XM_005250445.3 XP_005250502.1 7 - 8 848 145
XM_006716028.3 XP_006716091.1 7 - 8 848 145
XM_006716029.3 XP_006716092.1 7 - 8 848 145
XM_011516318.1 XP_011514620.1 8 - 9 892 145
XM_011516319.2 XP_011514621.1 8 - 9 976 145
XM_011516320.2 XP_011514622.1 7 - 8 848 145
XM_011516321.2 XP_011514623.1 7 - 8 848 145
XM_011516322.1 XP_011514624.1 8 - 9 892 145
XM_011516323.1 XP_011514625.1 8 - 9 892 145
XM_011516324.1 XP_011514626.1 8 - 9 564 145
XM_017012329.1 XP_016867818.1 9 - 10 943 145
XM_017012330.1 XP_016867819.1 9 - 10 943 145
XM_017012331.1 XP_016867820.1 8 - 9 973 145
XM_017012332.1 XP_016867821.1 8 - 9 892 145
XM_017012333.1 XP_016867822.1 8 - 9 892 145
XM_017012334.1 XP_016867823.1 9 - 10 943 145
XM_017012335.1 XP_016867824.1 9 - 10 943 145
XM_017012336.1 XP_016867825.1 8 - 9 892 145
XM_017012337.1 XP_016867826.1 9 - 10 943 145
XM_017012338.1 XP_016867827.1 8 - 9 971 145
XM_017012339.1 XP_016867828.1 9 - 10 943 145
XM_017012341.1 XP_016867830.1 7 - 8 589 145
GenBank mRNA AB009671.1 - 8 - 9 798 145
AF073291.2 - 8 - 9 723 145
AF155381.1 - 8 - 9 723 145
AF155382.1 - 8 - 9 723 145
AF158637.1 - 7 - 8 560 145
AK125432.1 - 7 - 8 863 145
AK291010.1 - 8 - 9 847 145
BC036029.1 - 8 - 9 892 145
BC092465.1 - 8 - 9 892 145
BC125118.1 - 8 - 9 754 145

View Full Product Details