The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184785.1 | NP_001171714.1 | 14 - 15 | 2400 | 103 |
NM_001184786.1 | NP_001171715.1 | 13 - 14 | 2361 | 103 | |
NM_001184787.1 | NP_001171716.1 | 14 - 15 | 2400 | 103 | |
NM_001184788.1 | NP_001171717.1 | 13 - 14 | 2361 | 103 | |
NM_001184789.1 | NP_001171718.1 | 13 - 14 | 2361 | 103 | |
NM_001184790.1 | NP_001171719.1 | 12 - 13 | 2229 | 103 | |
NM_001184791.1 | NP_001171720.1 | 12 - 13 | 2229 | 103 | |
NM_001184792.1 | NP_001171721.1 | 14 - 15 | 2400 | 103 | |
NM_001184793.1 | NP_001171722.1 | 14 - 15 | 2400 | 103 | |
NM_001184794.1 | NP_001171723.1 | 13 - 14 | 2361 | 103 | |
NM_019619.3 | NP_062565.2 | 14 - 15 | 2400 | 103 | |
XM_005252528.4 | XP_005252585.1 | 14 - 15 | 2391 | 103 | |
XM_005252530.4 | XP_005252587.1 | 14 - 15 | 2391 | 103 | |
XM_005252531.4 | XP_005252588.1 | 14 - 15 | 2391 | 103 | |
XM_005252532.4 | XP_005252589.1 | 13 - 14 | 2045 | 103 | |
XM_005252534.4 | XP_005252591.1 | 14 - 15 | 2391 | 103 | |
XM_005252535.4 | XP_005252592.1 | 13 - 14 | 2352 | 103 | |
XM_005252536.4 | XP_005252593.1 | 14 - 15 | 2391 | 103 | |
XM_011519569.2 | XP_011517871.1 | 14 - 15 | 2391 | 103 | |
XM_011519570.2 | XP_011517872.1 | 13 - 14 | 2352 | 103 | |
XM_011519571.2 | XP_011517873.1 | 14 - 15 | 2391 | 103 | |
XM_011519572.2 | XP_011517874.1 | 14 - 15 | 2391 | 103 | |
XM_011519573.2 | XP_011517875.1 | 14 - 15 | 2391 | 103 | |
XM_011519575.2 | XP_011517877.1 | 13 - 14 | 2075 | 103 | |
XM_011519576.2 | XP_011517878.1 | 13 - 14 | 2046 | 103 | |
XM_011519577.2 | XP_011517879.1 | 12 - 13 | 2223 | 103 | |
XM_011519578.2 | XP_011517880.1 | 12 - 13 | 2224 | 103 | |
XM_011519579.2 | XP_011517881.1 | 12 - 13 | 1944 | 103 | |
XM_011519581.2 | XP_011517883.1 | 14 - 15 | 2391 | 103 | |
XM_011519583.2 | XP_011517885.1 | 14 - 15 | 2391 | 103 | |
XM_011519584.2 | XP_011517886.1 | 14 - 15 | 2391 | 103 | |
XM_017016430.1 | XP_016871919.1 | 14 - 15 | 2391 | 103 | |
GenBank mRNA | AB073671.1 | - | 12 - 13 | 1899 | 103 |
AF177228.1 | - | 7 - 8 | 1136 | 103 | |
AF196185.1 | - | 14 - 15 | 2357 | 103 | |
AF196186.1 | - | 14 - 15 | 2357 | 103 | |
AF252293.1 | - | 12 - 13 | 1899 | 103 | |
AF332592.1 | - | 13 - 14 | 2031 | 103 | |
AF332593.1 | - | 13 - 14 | 2031 | 103 | |
AF454057.1 | - | 12 - 13 | 1777 | 103 | |
AF454058.1 | - | 12 - 13 | 2243 | 103 | |
AF454059.1 | - | 9 - 10 | 1364 | 103 | |
AF467002.1 | - | 14 - 15 | 2070 | 103 | |
AF467003.1 | - | 14 - 15 | 2070 | 103 | |
AF467004.1 | - | 14 - 15 | 2070 | 103 | |
AF467005.1 | - | 12 - 13 | 1899 | 103 | |
AF467006.1 | - | 13 - 14 | 2031 | 103 | |
AK024668.1 | - | 1 - 2 | 52 | 103 | |
AK025892.1 | - | 5 - 6 | 847 | 103 | |
AK027735.1 | - | 12 - 13 | 1697 | 103 | |
BC071566.1 | - | 13 - 14 | 2308 | 103 |