The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123226.1 | NP_001116698.1 | 4 - 5 | 950 | 85 |
NM_012123.3 | NP_036255.2 | 4 - 5 | 950 | 85 | |
NM_133645.2 | NP_598400.1 | 4 - 5 | 950 | 85 | |
XM_006715444.2 | XP_006715507.1 | 4 - 5 | 847 | 85 | |
XM_006715445.2 | XP_006715508.1 | 4 - 5 | 652 | 85 | |
XM_006715446.3 | XP_006715509.1 | 4 - 5 | 950 | 85 | |
XM_011535723.1 | XP_011534025.1 | 5 - 6 | 1039 | 85 | |
XM_011535724.1 | XP_011534026.1 | 5 - 6 | 907 | 85 | |
XM_011535725.2 | XP_011534027.1 | 5 - 6 | 775 | 85 | |
XM_017010697.1 | XP_016866186.1 | 4 - 5 | 847 | 85 | |
XM_017010698.1 | XP_016866187.1 | 5 - 6 | 1039 | 85 | |
GenBank mRNA | AF132937.1 | - | 4 - 5 | 872 | 85 |
AF319422.2 | - | 4 - 5 | 916 | 85 | |
AF469111.1 | - | 4 - 5 | 916 | 85 | |
AK074625.1 | - | 4 - 5 | 929 | 85 | |
AK225828.1 | - | 4 - 5 | 934 | 85 | |
AL833823.1 | - | 4 - 5 | 794 | 85 | |
AY078985.1 | - | 4 - 5 | 916 | 85 | |
AY078986.1 | - | 4 - 5 | 916 | 85 | |
BC005808.2 | - | 4 - 5 | 635 | 85 | |
BC011051.1 | - | 4 - 5 | 891 | 85 |