Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YLPM1
Entrez Gene ID
56252
Gene Name
YLP motif containing 1
Gene Aliases
C14orf170, PPP1R169, ZAP113, ZAP3
Chromosome Location
Chr.14: 74763322 - 74837310 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019589.2 NP_062535.2 2 - 3 1236 94
XM_005267860.3 XP_005267917.1 2 - 3 1280 94
XM_011536966.2 XP_011535268.1 3 - 4 395 94
XM_011536967.1 XP_011535269.1 2 - 3 1280 94
GenBank mRNA AK095760.1 - 2 - 3 1236 94
BC044636.1 - 2 - 3 597 94
DB096717.1 - 1 - 2 222 94

View Full Product Details