The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019589.2 | NP_062535.2 | 8 - 9 | 5153 | 89 |
XM_005267860.3 | XP_005267917.1 | 8 - 9 | 5197 | 89 | |
XM_011536966.2 | XP_011535268.1 | 9 - 10 | 4312 | 89 | |
XM_011536967.1 | XP_011535269.1 | 7 - 8 | 3079 | 89 | |
GenBank mRNA | AK090435.1 | - | 6 - 7 | 3909 | 89 |
AK297582.1 | - | 4 - 5 | 724 | 89 | |
AL832365.1 | - | 5 - 6 | 3688 | 89 | |
L40403.1 | - | 2 - 3 | 256 | 89 |