Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHOT2
Entrez Gene ID
89941
Gene Name
ras homolog family member T2
Gene Aliases
ARHT2, C16orf39, MIRO-2, MIRO2, RASL
Chromosome Location
Chr.16: 668083 - 674174 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138769.2 NP_620124.1 12 - 13 1072 88
XM_005255660.2 XP_005255717.1 12 - 13 1026 88
XM_005255661.2 XP_005255718.1 11 - 12 972 88
XM_005255662.2 XP_005255719.1 11 - 12 969 88
XM_005255663.2 XP_005255720.1 11 - 12 924 88
XM_006720970.2 XP_006721033.1 12 - 13 1009 88
XM_006720973.2 XP_006721036.1 11 - 12 907 88
XM_017023825.1 XP_016879314.1 12 - 13 1026 88
XM_017023826.1 XP_016879315.1 12 - 13 1023 88
XM_017023827.1 XP_016879316.1 11 - 12 972 88
XM_017023828.1 XP_016879317.1 11 - 12 969 88
XM_017023829.1 XP_016879318.1 11 - 12 924 88
XM_017023830.1 XP_016879319.1 10 - 11 870 88
XM_017023831.1 XP_016879320.1 12 - 13 1009 88
XM_017023832.1 XP_016879321.1 9 - 10 770 88
XM_017023833.1 XP_016879322.1 12 - 13 1006 88
XM_017023834.1 XP_016879323.1 10 - 11 816 88
XM_017023835.1 XP_016879324.1 12 - 13 1021 88
XM_017023836.1 XP_016879325.1 11 - 12 907 88
XM_017023837.1 XP_016879326.1 12 - 13 1006 88
XM_017023838.1 XP_016879327.1 11 - 12 887 88
XM_017023839.1 XP_016879328.1 12 - 13 1021 88
XM_017023840.1 XP_016879329.1 11 - 12 1035 88
XM_017023841.1 XP_016879330.1 10 - 11 1113 88
XM_017023842.1 XP_016879331.1 11 - 12 1078 88
GenBank mRNA AJ517413.1 - 12 - 13 955 88
AK024450.1 - 8 - 9 2502 88
AK090426.1 - 7 - 8 1356 88
AL834163.1 - 11 - 12 1090 88
AY207375.1 - 12 - 13 1069 88
BC014942.1 - 12 - 13 1022 88

View Full Product Details