Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHOT2
Entrez Gene ID
89941
Gene Name
ras homolog family member T2
Gene Aliases
ARHT2, C16orf39, MIRO-2, MIRO2, RASL
Chromosome Location
Chr.16: 668083 - 674174 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138769.2 NP_620124.1 10 - 11 867 92
XM_005255660.2 XP_005255717.1 10 - 11 821 92
XM_005255661.2 XP_005255718.1 9 - 10 767 92
XM_005255662.2 XP_005255719.1 9 - 10 764 92
XM_005255663.2 XP_005255720.1 9 - 10 719 92
XM_006720970.2 XP_006721033.1 10 - 11 804 92
XM_006720973.2 XP_006721036.1 9 - 10 702 92
XM_017023825.1 XP_016879314.1 10 - 11 821 92
XM_017023826.1 XP_016879315.1 10 - 11 818 92
XM_017023827.1 XP_016879316.1 9 - 10 767 92
XM_017023828.1 XP_016879317.1 9 - 10 764 92
XM_017023829.1 XP_016879318.1 9 - 10 719 92
XM_017023830.1 XP_016879319.1 8 - 9 665 92
XM_017023831.1 XP_016879320.1 10 - 11 804 92
XM_017023832.1 XP_016879321.1 7 - 8 565 92
XM_017023833.1 XP_016879322.1 10 - 11 801 92
XM_017023834.1 XP_016879323.1 8 - 9 611 92
XM_017023835.1 XP_016879324.1 10 - 11 816 92
XM_017023836.1 XP_016879325.1 9 - 10 702 92
XM_017023837.1 XP_016879326.1 10 - 11 801 92
XM_017023838.1 XP_016879327.1 9 - 10 682 92
XM_017023839.1 XP_016879328.1 10 - 11 816 92
GenBank mRNA AJ517413.1 - 10 - 11 750 92
AL834163.1 - 9 - 10 885 92
AY207375.1 - 10 - 11 864 92
BC014942.1 - 10 - 11 817 92

View Full Product Details