The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145102.1 | NP_001138574.1 | 2 - 3 | 463 | 65 |
NM_001145103.1 | NP_001138575.1 | 2 - 3 | 274 | 65 | |
NM_005902.3 | NP_005893.1 | 2 - 3 | 698 | 65 | |
XM_011521560.1 | XP_011519862.1 | 2 - 3 | 637 | 65 | |
GenBank mRNA | AB209616.1 | - | 2 - 3 | 283 | 65 |
AK290881.1 | - | 3 - 4 | 623 | 65 | |
AK298139.1 | - | 2 - 3 | 274 | 65 | |
AK316017.1 | - | 2 - 3 | 323 | 65 | |
BC050743.1 | - | 2 - 3 | 616 | 65 | |
DA453132.1 | - | 2 - 3 | 463 | 65 | |
U68019.1 | - | 2 - 3 | 466 | 65 | |
U76622.1 | - | 2 - 3 | 437 | 65 |