The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005932.3 | NP_005923.2 | 10 - 11 | 1228 | 64 |
XM_011535097.2 | XP_011533399.1 | 10 - 11 | 991 | 64 | |
XM_011535098.2 | XP_011533400.1 | 10 - 11 | 1228 | 64 | |
GenBank mRNA | AK225290.1 | - | 10 - 11 | 1178 | 64 |
AK291923.1 | - | 10 - 11 | 1104 | 64 | |
BC009934.2 | - | 10 - 11 | 1200 | 64 | |
U80034.1 | - | 10 - 11 | 1174 | 64 |