The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014423.3 | NP_055238.1 | 1 - 2 | 410 | 75 |
XM_006714587.3 | XP_006714650.1 | 1 - 2 | 410 | 75 | |
GenBank mRNA | AB209547.1 | - | 1 - 2 | 366 | 75 |
AF197927.1 | - | 1 - 2 | 339 | 75 | |
AF213987.2 | - | 1 - 2 | 67 | 75 | |
AK055638.1 | - | 1 - 2 | 358 | 75 | |
AK307407.1 | - | 1 - 2 | 410 | 75 | |
BC025700.1 | - | 1 - 2 | 262 | 75 | |
BC063007.1 | - | 1 - 2 | 261 | 75 | |
BC100287.1 | - | 1 - 2 | 362 | 75 | |
BC137226.1 | - | 1 - 2 | 308 | 75 | |
BC144538.1 | - | 1 - 2 | 308 | 75 | |
DA934874.1 | - | 1 - 2 | 410 | 75 |