The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308006.1 | NP_001294935.1 | 6 - 7 | 1219 | 66 |
NM_006033.3 | NP_006024.1 | 7 - 8 | 1441 | 66 | |
XM_005258390.1 | XP_005258447.1 | 7 - 8 | 2258 | 66 | |
XM_011526265.2 | XP_011524567.1 | 6 - 7 | 1249 | 66 | |
XM_011526267.1 | XP_011524569.1 | 7 - 8 | 1828 | 66 | |
XM_017026095.1 | XP_016881584.1 | 7 - 8 | 1940 | 66 | |
GenBank mRNA | AF118767.1 | - | 7 - 8 | 1415 | 66 |
AK291799.1 | - | 7 - 8 | 1415 | 66 | |
AK300333.1 | - | 6 - 7 | 1190 | 66 | |
AK315252.1 | - | 7 - 8 | 1443 | 66 | |
BC060825.1 | - | 7 - 8 | 1412 | 66 |