The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184856.1 | NP_001171785.1 | 10 - 11 | 1574 | 90 |
NM_017566.3 | NP_060036.2 | 11 - 12 | 1667 | 90 | |
XM_005255999.1 | XP_005256056.1 | 10 - 11 | 1587 | 90 | |
XM_006721201.1 | XP_006721264.1 | 11 - 12 | 1687 | 90 | |
GenBank mRNA | AK023986.1 | - | 11 - 12 | 1624 | 90 |
AK057211.1 | - | 2 - 3 | 1813 | 90 | |
AK098630.1 | - | 7 - 8 | 1115 | 90 | |
BC011680.2 | - | 10 - 11 | 1558 | 90 | |
BC022969.1 | - | 11 - 12 | 1611 | 90 |