Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHDC4
Entrez Gene ID
54758
Gene Name
kelch domain containing 4
Gene Aliases
-
Chromosome Location
Chr.16: 87693537 - 87765997 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184854.1 NP_001171783.1 1 - 2 200 101
GenBank mRNA BC001044.2 - 1 - 2 107 101

View Full Product Details