The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014443.2 | NP_001014443.1 | 12 - 13 | 1867 | 81 |
NM_012475.4 | NP_036607.3 | 11 - 12 | 1726 | 81 | |
XM_017001036.1 | XP_016856525.1 | 10 - 11 | 2011 | 81 | |
XM_017001037.1 | XP_016856526.1 | 11 - 12 | 2065 | 81 | |
XM_017001038.1 | XP_016856527.1 | 11 - 12 | 2101 | 81 | |
XM_017001039.1 | XP_016856528.1 | 10 - 11 | 1954 | 81 | |
XM_017001040.1 | XP_016856529.1 | 11 - 12 | 1789 | 81 | |
XM_017001041.1 | XP_016856530.1 | 11 - 12 | 1750 | 81 | |
XM_017001042.1 | XP_016856531.1 | 6 - 7 | 909 | 81 | |
GenBank mRNA | AB208899.1 | - | 7 - 8 | 2643 | 81 |
AF177758.1 | - | 11 - 12 | 1577 | 81 | |
AF233442.1 | - | 11 - 12 | 1664 | 81 | |
AK292319.1 | - | 12 - 13 | 1812 | 81 | |
AK310326.1 | - | 11 - 12 | 1759 | 81 | |
AL157417.1 | - | 7 - 8 | 2042 | 81 | |
BC003130.1 | - | 10 - 11 | 1226 | 81 | |
BC090946.1 | - | 12 - 13 | 1815 | 81 | |
BC136291.1 | - | 10 - 11 | 1514 | 81 |