Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HES4
Entrez Gene ID
57801
Gene Name
hes family bHLH transcription factor 4
Gene Aliases
bHLHb42
Chromosome Location
Chr.1: 998962 - 1001052 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142467.1 NP_001135939.1 1 - 2 484 56
NM_021170.3 NP_066993.1 2 - 3 406 56
GenBank mRNA AB048791.1 - 2 - 3 207 56
BC012351.1 - 2 - 3 324 56
BI757407.1 - 1 - 2 484 56

View Full Product Details