Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRIQ1
Entrez Gene ID
84125
Gene Name
leucine rich repeats and IQ motif containing 1
Gene Aliases
-
Chromosome Location
Chr.12: 85036321 - 85273004 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079910.1 NP_001073379.1 18 - 19 4323 75
XM_011538817.2 XP_011537119.1 18 - 19 4266 75
XM_011538818.2 XP_011537120.1 18 - 19 4266 75
XM_011538819.2 XP_011537121.1 18 - 19 4266 75
XM_011538823.2 XP_011537125.1 18 - 19 4266 75
XM_011538824.2 XP_011537126.1 18 - 19 4266 75
XM_017020019.1 XP_016875508.1 18 - 19 4266 75
GenBank mRNA CA502842.1 - 2 - 3 250 75
DN831755.1 - 3 - 4 694 75

View Full Product Details