The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079910.1 | NP_001073379.1 | 2 - 3 | 249 | 130 |
XM_011538817.2 | XP_011537119.1 | 2 - 3 | 192 | 130 | |
XM_011538818.2 | XP_011537120.1 | 2 - 3 | 192 | 130 | |
XM_011538819.2 | XP_011537121.1 | 2 - 3 | 192 | 130 | |
XM_011538823.2 | XP_011537125.1 | 2 - 3 | 192 | 130 | |
XM_011538824.2 | XP_011537126.1 | 2 - 3 | 192 | 130 | |
XM_011538825.2 | XP_011537127.1 | 2 - 3 | 192 | 130 | |
XM_017020019.1 | XP_016875508.1 | 2 - 3 | 192 | 130 | |
GenBank mRNA | AK022365.1 | - | 2 - 3 | 250 | 130 |
AK307799.1 | - | 2 - 3 | 199 | 130 | |
AU122206.2 | - | 2 - 3 | 250 | 130 | |
BC093088.1 | - | 2 - 3 | 220 | 130 | |
DB447399.2 | - | 2 - 3 | 259 | 130 |