Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRSS23
Entrez Gene ID
11098
Gene Name
protease, serine 23
Gene Aliases
SIG13, SPUVE, ZSIG13
Chromosome Location
Chr.11: 86791059 - 86952910 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293178.1 NP_001280107.1 3 - 3 3317 141
NM_001293179.1 NP_001280108.1 2 - 2 3408 141
NM_001293180.1 NP_001280109.1 2 - 2 3138 141
NM_007173.5 NP_009104.2 2 - 2 3413 141
GenBank mRNA AK075362.1 - 2 - 2 3109 141
BG286404.1 - 1 - 1 471 137

View Full Product Details