Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSE1
Entrez Gene ID
23199
Gene Name
Gse1 coiled-coil protein
Gene Aliases
CRHSP24, KIAA0182
Chromosome Location
Chr.16: 85169512 - 85676206 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134473.2 NP_001127945.1 12 - 13 3014 77
NM_001278184.1 NP_001265113.1 12 - 13 3068 77
NM_014615.3 NP_055430.1 13 - 14 3287 77
XM_005255859.4 XP_005255916.3 14 - 15 5429 77
XM_005255860.3 XP_005255917.3 13 - 14 5322 77
XM_005255861.4 XP_005255918.3 13 - 14 5210 77
XM_005255863.4 XP_005255920.1 13 - 14 3179 77
XM_005255864.4 XP_005255921.1 13 - 14 3911 77
XM_005255865.4 XP_005255922.1 12 - 13 3692 77
XM_011522965.2 XP_011521267.1 13 - 14 3678 77
XM_011522966.2 XP_011521268.1 13 - 14 3145 77
XM_017023083.1 XP_016878572.1 13 - 14 3437 77
GenBank mRNA AK127344.1 - 2 - 3 1698 77
AK308612.1 - 3 - 4 736 77
BC000753.2 - 1 - 2 256 77
BC037556.1 - 12 - 13 3037 77
D80004.2 - 12 - 13 2952 77

View Full Product Details