The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134473.2 | NP_001127945.1 | 7 - 8 | 1526 | 157 |
NM_001278184.1 | NP_001265113.1 | 7 - 8 | 1580 | 157 | |
NM_014615.3 | NP_055430.1 | 8 - 9 | 1799 | 157 | |
XM_005255859.4 | XP_005255916.3 | 9 - 10 | 3941 | 157 | |
XM_005255860.3 | XP_005255917.3 | 9 - 10 | 3948 | 157 | |
XM_005255861.4 | XP_005255918.3 | 8 - 9 | 3722 | 157 | |
XM_005255863.4 | XP_005255920.1 | 8 - 9 | 1691 | 157 | |
XM_005255864.4 | XP_005255921.1 | 8 - 9 | 2423 | 157 | |
XM_005255865.4 | XP_005255922.1 | 7 - 8 | 2204 | 157 | |
XM_011522965.2 | XP_011521267.1 | 8 - 9 | 2190 | 157 | |
XM_011522966.2 | XP_011521268.1 | 8 - 9 | 1657 | 157 | |
XM_017023083.1 | XP_016878572.1 | 8 - 9 | 1949 | 157 | |
GenBank mRNA | BC037556.1 | - | 7 - 8 | 1549 | 157 |
D80004.2 | - | 7 - 8 | 1464 | 157 |