The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288713.1 | NP_001275642.1 | 12 - 13 | 1425 | 93 |
NM_001288714.1 | NP_001275643.1 | 10 - 11 | 1170 | 93 | |
NM_006157.4 | NP_006148.2 | 11 - 12 | 1341 | 93 | |
NM_201551.2 | NP_963845.1 | 11 - 12 | 1341 | 93 | |
GenBank mRNA | AK127805.1 | - | 12 - 13 | 1405 | 93 |
AK299558.1 | - | 8 - 9 | 883 | 93 | |
AK303736.1 | - | 10 - 11 | 1139 | 93 | |
AK313445.1 | - | 11 - 12 | 1321 | 93 | |
BC069674.1 | - | 11 - 12 | 1232 | 93 | |
BC096100.1 | - | 11 - 12 | 1231 | 93 | |
BC096101.1 | - | 11 - 12 | 1231 | 93 | |
BC096102.3 | - | 11 - 12 | 1231 | 93 | |
BC096103.3 | - | 11 - 12 | 1231 | 93 | |
D83017.1 | - | 11 - 12 | 1270 | 93 | |
JX565026.1 | - | 10 - 11 | 1049 | 93 |