Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLLT3
Entrez Gene ID
4300
Gene Name
MLLT3, super elongation complex subunit
Gene Aliases
AF9, YEATS3
Chromosome Location
Chr.9: 20341665 - 20622543 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286691.1 NP_001273620.1 6 - 7 1404 104
NM_004529.3 NP_004520.2 6 - 7 1488 104
XM_017014726.1 XP_016870215.1 6 - 7 1403 104
XM_017014727.1 XP_016870216.1 6 - 7 1399 104
GenBank mRNA AK225957.1 - 6 - 7 1420 104
AK297587.1 - 5 - 6 1515 104
AK301474.1 - 6 - 7 1290 104
BC036089.1 - 6 - 7 1451 104
BX649194.1 - 6 - 7 1464 104
D16688.1 - 2 - 3 524 104
KU178189.1 - 5 - 6 1201 104
L13744.1 - 6 - 7 1396 104

View Full Product Details