Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCF4
Entrez Gene ID
6925
Gene Name
transcription factor 4
Gene Aliases
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location
Chr.18: 55222331 - 55635993 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083962.1 NP_001077431.1 14 - 15 1759 114
NM_001243226.2 NP_001230155.2 15 - 16 1549 114
NM_001243227.1 NP_001230156.1 13 - 14 1593 114
NM_001243228.1 NP_001230157.1 14 - 15 1703 114
NM_001243230.1 NP_001230159.1 13 - 14 1232 114
NM_001243231.1 NP_001230160.1 12 - 13 1193 114
NM_001243232.1 NP_001230161.1 10 - 11 1215 114
NM_001243233.1 NP_001230162.1 10 - 11 1156 114
NM_001243234.1 NP_001230163.1 7 - 8 788 114
NM_001243235.1 NP_001230164.1 7 - 8 788 114
NM_001243236.1 NP_001230165.1 7 - 8 804 114
NM_001306207.1 NP_001293136.1 13 - 14 1384 114
NM_001306208.1 NP_001293137.1 10 - 11 1215 114
NM_003199.2 NP_003190.1 14 - 15 1759 114
XM_005266739.3 XP_005266796.2 13 - 14 1399 114
XM_005266741.3 XP_005266798.1 14 - 15 1359 114
XM_005266743.4 XP_005266800.1 13 - 14 1220 114
XM_005266744.3 XP_005266801.1 13 - 14 1340 114
XM_005266745.3 XP_005266802.1 13 - 14 1202 114
XM_005266747.3 XP_005266804.1 12 - 13 1196 114
XM_005266749.3 XP_005266806.1 10 - 11 987 114
XM_005266752.4 XP_005266809.1 10 - 11 1557 114
XM_005266754.3 XP_005266811.1 9 - 10 981 114
XM_005266755.4 XP_005266812.1 10 - 11 1016 114
XM_005266761.3 XP_005266818.1 7 - 8 1297 114
XM_006722536.2 XP_006722599.1 14 - 15 1682 114
XM_006722537.2 XP_006722600.1 14 - 15 1315 114
XM_006722538.2 XP_006722601.1 13 - 14 1337 114
XM_011526158.2 XP_011524460.1 13 - 14 2313 114
XM_011526160.2 XP_011524462.1 9 - 10 1366 114
XM_017025934.1 XP_016881423.1 13 - 14 1396 114
XM_017025935.1 XP_016881424.1 13 - 14 1396 114
XM_017025936.1 XP_016881425.1 13 - 14 1399 114
XM_017025937.1 XP_016881426.1 13 - 14 1153 114
XM_017025938.1 XP_016881427.1 14 - 15 1312 114
XM_017025939.1 XP_016881428.1 14 - 15 1359 114
XM_017025940.1 XP_016881429.1 14 - 15 1682 114
XM_017025941.1 XP_016881430.1 14 - 15 1315 114
XM_017025942.1 XP_016881431.1 14 - 15 1359 114
XM_017025943.1 XP_016881432.1 14 - 15 1359 114
XM_017025944.1 XP_016881433.1 13 - 14 1617 114
XM_017025945.1 XP_016881434.1 13 - 14 1617 114
XM_017025946.1 XP_016881435.1 13 - 14 1634 114
XM_017025947.1 XP_016881436.1 13 - 14 1220 114
XM_017025948.1 XP_016881437.1 13 - 14 1617 114
XM_017025949.1 XP_016881438.1 13 - 14 1337 114
XM_017025950.1 XP_016881439.1 13 - 14 1614 114
XM_017025951.1 XP_016881440.1 12 - 13 1193 114
XM_017025952.1 XP_016881441.1 10 - 11 1207 114
XM_017025953.1 XP_016881442.1 10 - 11 987 114
XM_017025954.1 XP_016881443.1 10 - 11 1568 114
XM_017025955.1 XP_016881444.1 9 - 10 1302 114
XM_017025956.1 XP_016881445.1 9 - 10 981 114
XM_017025957.1 XP_016881446.1 7 - 8 1297 114
GenBank mRNA AB209741.1 - 9 - 10 867 114
AK095041.1 - 10 - 11 1215 114
AK096862.1 - 12 - 13 1193 114
AK122765.1 - 14 - 15 1759 114
AK299169.1 - 13 - 14 1593 114
AK300038.1 - 7 - 8 734 114
AK300612.1 - 10 - 11 1156 114
AK300636.1 - 9 - 10 886 114
AK301144.1 - 13 - 14 1370 114
AK315074.1 - 15 - 16 1691 114
AK316165.1 - 9 - 10 930 114
AL518903.3 - 7 - 8 804 114
BC031056.1 - 15 - 16 1510 114
BC125084.1 - 13 - 14 1173 114
BC125085.1 - 13 - 14 1173 114
FR748210.1 - 13 - 14 1147 114
FR748211.1 - 13 - 14 1147 114
FR748212.1 - 11 - 12 967 114
FR748213.1 - 11 - 12 967 114
FR748214.1 - 12 - 13 1075 114
FR748215.1 - 10 - 11 895 114
FR748216.1 - 13 - 14 1138 114
FR748217.1 - 12 - 13 1021 114
FR748218.1 - 8 - 9 757 114
FR748219.1 - 10 - 11 934 114
FR748220.1 - 7 - 8 667 114
FR748221.1 - 7 - 8 667 114
FR748222.1 - 7 - 8 667 114
FR748223.1 - 5 - 6 499 114
M74718.1 - 7 - 8 619 114
M74719.1 - 14 - 15 1346 114
X52079.1 - 11 - 12 1012 114

View Full Product Details