Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX29
Entrez Gene ID
92017
Gene Name
sorting nexin 29
Gene Aliases
A-388D4.1, RUNDC2A
Chromosome Location
Chr.16: 11976734 - 12574289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032167.4 NP_115543.3 13 - 14 1666 88
XM_005255682.3 XP_005255739.1 13 - 14 1691 88
XM_011522738.2 XP_011521040.1 13 - 14 1838 88
XM_011522741.2 XP_011521043.1 13 - 14 1822 88
XM_011522743.2 XP_011521045.1 13 - 14 1826 88
XM_011522744.2 XP_011521046.1 13 - 14 1697 88
XM_017023871.1 XP_016879360.1 13 - 14 1835 88
XM_017023872.1 XP_016879361.1 13 - 14 1655 88
XM_017023873.1 XP_016879362.1 13 - 14 1822 88
XM_017023874.1 XP_016879363.1 13 - 14 1655 88
XM_017023875.1 XP_016879364.1 11 - 12 1362 88
XM_017023878.1 XP_016879367.1 3 - 4 1160 88
GenBank mRNA AK074072.1 - 3 - 4 273 88
BC029857.1 - 1 - 2 60 88

View Full Product Details