The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032167.4 | NP_115543.3 | 14 - 15 | 1754 | 87 |
XM_005255682.3 | XP_005255739.1 | 14 - 15 | 1779 | 87 | |
XM_011522738.2 | XP_011521040.1 | 14 - 15 | 1926 | 87 | |
XM_011522741.2 | XP_011521043.1 | 14 - 15 | 1910 | 87 | |
XM_011522743.2 | XP_011521045.1 | 14 - 15 | 1914 | 87 | |
XM_011522744.2 | XP_011521046.1 | 14 - 15 | 1785 | 87 | |
XM_017023871.1 | XP_016879360.1 | 14 - 15 | 1923 | 87 | |
XM_017023872.1 | XP_016879361.1 | 14 - 15 | 1743 | 87 | |
XM_017023873.1 | XP_016879362.1 | 14 - 15 | 1910 | 87 | |
XM_017023874.1 | XP_016879363.1 | 14 - 15 | 1743 | 87 | |
XM_017023875.1 | XP_016879364.1 | 12 - 13 | 1450 | 87 | |
XM_017023878.1 | XP_016879367.1 | 4 - 5 | 1248 | 87 | |
GenBank mRNA | AK074072.1 | - | 4 - 5 | 361 | 87 |
BC029857.1 | - | 2 - 3 | 148 | 87 |