The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286503.1 | NP_001273432.1 | 10 - 11 | 1817 | 145 |
NM_001286504.1 | NP_001273433.1 | 10 - 11 | 1678 | 145 | |
NM_001286505.1 | NP_001273434.1 | 8 - 9 | 1444 | 145 | |
NM_006644.3 | NP_006635.2 | 10 - 11 | 1817 | 145 | |
XM_005266236.1 | XP_005266293.1 | 10 - 11 | 2399 | 145 | |
XM_011534887.2 | XP_011533189.1 | 10 - 11 | 2399 | 145 | |
XM_011534888.1 | XP_011533190.1 | 8 - 9 | 1507 | 145 | |
XM_017020361.1 | XP_016875850.1 | 10 - 11 | 1741 | 145 | |
XM_017020362.1 | XP_016875851.1 | 10 - 11 | 2399 | 145 | |
XM_017020363.1 | XP_016875852.1 | 10 - 11 | 1741 | 145 | |
XM_017020364.1 | XP_016875853.1 | 8 - 9 | 1507 | 145 | |
GenBank mRNA | AB003333.1 | - | 10 - 11 | 1691 | 145 |
AB003334.1 | - | 10 - 11 | 1723 | 145 | |
AF039695.1 | - | 10 - 11 | 1420 | 145 | |
AK293955.1 | - | 10 - 11 | 1693 | 145 | |
AK302294.1 | - | 8 - 9 | 1095 | 145 | |
AK302430.1 | - | 10 - 11 | 1678 | 145 | |
AK302832.1 | - | 12 - 13 | 1768 | 145 | |
AK314576.1 | - | 10 - 11 | 1741 | 145 | |
BC037553.1 | - | 10 - 11 | 1719 | 145 | |
BX648623.1 | - | 4 - 5 | 2025 | 145 | |
D86956.1 | - | 10 - 11 | 1725 | 145 |