The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007249.4 | NP_009180.3 | 3 - 4 | 351 | 78 |
XM_005266251.3 | XP_005266308.1 | 3 - 4 | 460 | 78 | |
XM_011534907.2 | XP_011533209.1 | 2 - 3 | 174 | 78 | |
XM_011534908.2 | XP_011533210.1 | 2 - 3 | 174 | 78 | |
XM_011534909.2 | XP_011533211.1 | 3 - 4 | 307 | 78 | |
XM_011534910.2 | XP_011533212.1 | 2 - 3 | 138 | 78 | |
XM_011534911.2 | XP_011533213.1 | 2 - 3 | 216 | 78 | |
XM_011534912.2 | XP_011533214.1 | 2 - 3 | 287 | 78 | |
XM_017020384.1 | XP_016875873.1 | 2 - 3 | 118 | 78 | |
GenBank mRNA | AF113122.1 | - | 3 - 4 | 325 | 78 |
AF161471.1 | - | 3 - 4 | 199 | 78 | |
AF330041.1 | - | 2 - 3 | 137 | 78 | |
AJ243274.1 | - | 2 - 3 | 129 | 78 | |
AK291397.1 | - | 3 - 4 | 447 | 78 | |
BC019680.1 | - | 2 - 3 | 156 | 78 | |
DB125574.1 | - | 3 - 4 | 196 | 78 | |
HF546212.1 | - | 3 - 4 | 322 | 78 |